Human recombinant netrin 1 (from HEK293 cells)
Fournisseur: ProSci
PRSI90-261EA 631 EUR InStock
PRSI90-261
Human recombinant netrin 1 (from HEK293 cells)
Protéines et Peptides
Netrin-1 controls guidance of CNS commissural axons and peripheral motor axons. Its association with either DCC or some UNC5 receptors will lead to axon attraction or repulsion, respectively. It also serve as a survival factor via its association with its receptors which prevent the initiation of apoptosis. Netrin 1 is also Involved in tumorigenesis by regulating apoptosis.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité