Human recombinant POP1 (from E. coli)
Fournisseur: ProSci
PRSI90-303EA 365 EUR InStock
PRSI90-303
Human recombinant POP1 (from E. coli)
Protéines et Peptides
The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin (IL)-1beta and IL-18. Members of the Nod-like receptor family (NLRs), including NLRP3/NALP3, are critical components of the inflammasome that link danger-signals to caspase-1 activation. Defects in NLRP3 are the cause of familial cold autoinflammatory syndrome type 1 (FCAS1), Muckle-Wells syndrome (MWS) and of chronic infantile neurologic cutaneous and articular syndrome (CINCA). Pyrin domain-containing protein 1 (POP1) interacts with the protein Asc (Pycard) and modulates the interaction of Asc to NLR proteins.
Fusion-Tag: GST tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité