Human recombinant Angiopoietin 2 (from HEK293 cells)
Fournisseur: ProSci
PRSI90-311EA 400 EUR InStock
PRSI90-311
Human recombinant Angiopoietin 2 (from HEK293 cells)
Protéines et Peptides
Angiopoietin-1 (Ang-1) and Angiopoietin-2 (Ang-2) are closely related secreted ligands which bind with similar affinity to Tie-2. Tie-2 and angiopoietins have been shown to play critical roles in embryogenic angiogenesis and in maintaining the integrity of the adult vasculature. Ang-1 activates Tie-2 signalling on endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth and stabilisation. Ang-2 has been identified as a secreted protein ligand of Tie-2 and has alternatively been reported to be an antagonist for Ang-1 induced Tie-2 signalling as well as an agonist for Tie-2 signalling, depending on the cell context.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité