Human recombinant FasL (soluble) (from HEK293 cells)
Fournisseur: ProSci
PRSI90-312EA 537 EUR InStock
PRSI90-312
Human recombinant FasL (soluble) (from HEK293 cells)
Protéines et Peptides
Fc (human):FasL, Soluble (human) is a high activity construct in which two trimeric FasL are artificially linked via the Fc binding domain of human IgG1. This construct very effectively simulates the natural membrane-assisted aggregation of FasL in vivo. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. It is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité