Human recombinant Frizzled-5 (from HEK293 cells)
Fournisseur: ProSci
PRSI90-313EA 537 EUR InStock
PRSI90-313
Human recombinant Frizzled-5 (from HEK293 cells)
Protéines et Peptides
Wnts are lipid-modified morphogens that control many processes during development, stem cell maintenance and homeostasis and their aberrant regulation has been linked to diseases in man including diabetes, neurodegeneration and cancer. Three distinct receptor families mediate Wnt signalling: Fzs, the single-pass LRP5/6 co-receptors and the atypical tyrosine kinase receptors Ror2 and Ryk. Frizzled proteins can activate canonical Wnt/beta-catenin signalling as well as non-canonical pathways (Wnt/Ca2+ pathways and planar cell polarity). The cysteine-rich domain (CRD) of the frizzled receptors situated on the extracellular part of the receptor is sufficient to bind the Wnt ligands.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité