Human recombinant FNDC5:Fc (from HEK293 cells)
Fournisseur: ProSci
PRSI90-334EA 471 EUR InStock
PRSI90-334
Human recombinant FNDC5:Fc (from HEK293 cells)
Protéines et Peptides
Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin). Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure. The membrane receptor FNDC5 can be cleaved by an unknown protease to release the Irisin protein (aa 32-143 for human) and potentially also the FNDC5 (extracellular domain, aa 32-153 for human).
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité