Human recombinant PARP3 (from Insect cells (Sf21))
Fournisseur: ProSci
PRSI90-342EA 506 EUR InStock
PRSI90-342
Human recombinant PARP3 (from Insect cells (Sf21))
Protéines et Peptides
PARP-3 [ARTD3] is involved in the base excision repair (BER) pathway, by catalyzing the poly(ADP-ribosyl)ation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signalling pathway leading to the reparation of DNA strand breaks. May link the DNA damage surveillance network to the mitotic fidelity checkpoint. Negatively influences the G1/S cell cycle progression without interfering with centrosome duplication. May be involved in the regulation of PRC2 and PRC3 complex-dependent gene silencing.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité