Human Recombinant CCL24 (from CHO cells)
Fournisseur: ProSci
PRSI90-389EA 471 EUR InStock
PRSI90-389
Human Recombinant CCL24 (from CHO cells)
Protéines et Peptides
CCL24 (eotaxin-2) has been shown to induce chemotaxis of eosinophils, basophils and resting T lymphocytes but not monocytes and activated T lymphocytes. CCL24 is a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. CCL24 interacts with chemokine receptor CCR3 to induce chemotaxis in eosinophils. Elevated levels of CCL24 has been seen in patients with aspirin-exacerbated respiratory disease (AERD), such as asthma.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité