Mouse recombinant CD275 non-lytic (from CHO cells)
Fournisseur: ProSci
PRSI90-437EA 584 EUR InStock
PRSI90-437
Mouse recombinant CD275 non-lytic (from CHO cells)
Protéines et Peptides
Mouse CD275 (B7-H2) is a member of the growing B7 family of immune costimulatory proteins. CD275 has been identified as the ligand for ICOS, a member of the CD28 family of co-stimulatory receptors. Mouse CD275 is expressed on resting B cells and at low levels on monocytes. The CD275/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité