Human recombinant CX3CL1 (from CHO cells)
Fournisseur: ProSci
PRSI90-453EA 612 EUR InStock
PRSI90-453
Human recombinant CX3CL1 (from CHO cells)
Protéines et Peptides
CX3CL1 is expressed on TNF-alpha and IL-1 activated endothelial cells (ECs) and binds to CX3CR1. CX3CL1 can exist in 2 forms, as membrane-anchored or as a shed 95-kDa glycoprotein. The soluble CX3CL1 has potent chemoattractant activity for T cells and monocytes, and the cell surface bound protein, which is induced on activated primary endothelial cells, promotes strong adhesion of those leukocytes. CX3CL1 may play a role in regulating leukocyte adhesion and migration processes at the endothelium.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité