Human recombinant IL6 Receptor (from CHO cells)
Fournisseur: ProSci
PRSI90-487EA 525 EUR InStock
PRSI90-487
Human recombinant IL6 Receptor (from CHO cells)
Protéines et Peptides
Interleukin-6 (IL-6) is a multifunctional factor, which exerts its activities through binding to a highaffinity receptor complex consisting of two membrane glycoproteins: an 80 kDa component receptor that binds IL-6 with low affinity (IL-6Ralpha) and a signal transducing component of 130 kDa (gp130) that does not bind IL-6 by itself, but is required for highaffinity binding of IL-6 by the complex. A soluble form of the IL-6Ralpha has been found in the urine of healthy adult humans.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité