Mouse recombinant SCF non-lytic (from CHO cells)
Fournisseur: ProSci
PRSI90-542EA 471 EUR InStock
PRSI90-542
Mouse recombinant SCF non-lytic (from CHO cells)
Protéines et Peptides
Stem cell factor (SCF), also known as cKit ligand (KL), mast cell growth factor (MGF), and steel factor (SLF), is a widely expressed 28-40 kDa type I transmembrane glycoprotein. It promotes the survival, differentiation, and mobilisaion of multiple cell types including myeloid, erythroid, megakaryocytic, lymphoid, germ cell, and melanocyte progenitors. SCF is a primary growth and activation factor for mast cells and eosinophils. Noncovalent dimers of transmembrane or soluble SCF interact with the receptor tyrosine kinase SCF R/cKit to trigger receptor dimerisaion and signalling. SCF assists in the recovery of cardiac function following myocardial infarction by increasing the number of cardiomyocytes and vascular channels.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité