Human recombinant Butyrylcholine Esterase (from Cells)
Fournisseur: ProSci
PRSI91-508EA 789 EUR InStock
PRSI91-508
Human recombinant Butyrylcholine Esterase (from Cells)
Enzymes
Butyrylcholine Esterase (BCHE) is a secreted protein that belongs to the type-B carboxylesterase/lipase family. BCHE is a major acetylcholine hydrolyzing enzyme in the circulation. It is detected in blood plasma and present in most cells except erythrocytes. BCHE is an esterase with broad substrate specificity. BCHE can contribute to the inactivation of the neurotransmitter acetylcholine. BCHE can degrade a large number of neurotoxic organophosphate esters. Thus, it plays important pharmacological and toxicological roles and is thought to be involved in the pathological progression. Defects in BCHE are the cause of butyrylcholinesterase deficiency (BChE deficiency) which is a metabolic disorder characterised by prolonged apnoea after the use of certain anesthetic drugs, including the muscle relaxants succinylcholine and other ester local anesthetics.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité