Human recombinant Wnt inhibitory factor 1 protein (from cells)
Fournisseur: ProSci
PRSI91-363EA 490 EUR InStock
PRSI91-363
Human recombinant Wnt inhibitory factor 1 protein (from cells)
Protéines et Peptides
Wnt Inhibitory Factor 1 (WIF1) is a secreted protein, which binds WNT proteins and inhibits their activities. WNT proteins are extracellular signalling molecules involved in the control of embryonic development. WIF1 contains a WNT inhibitory factor (WIF) domain and 5 epidermal growth factor (EGF)-like domains. is found to be present in fish, amphibia and mammals. WIF1 is a recurrent target in human salivary gland oncogenesis.WIF1 may be involved in mesoderm segmentation. WIF1 is a tumour suppressor, specifically in nonfunctioning pituitary tumours.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité