Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-GHSR Rabbit Polyclonal Antibody

Fournisseur: Enzo Life Sciences
ENZOADI905792100EA 442.2 EUR InStock
ENZOADI905792100 ALEXADI905792100
Anti-GHSR Rabbit Polyclonal Antibody
Anticorps
Ghrelin is a multifunctional peptide hormone secreted from the anterior pituitary known to affect many physiological processes including the release of growth hormone, appetite regulation, gastric acid secretion, gastrointestinal motility, and blood pressure regulation. The 28 amino acid ghrelin peptide is modified at Ser3 by addition of a fatty acid group (i.e., n-octanoyl) that is essential for its biological activity. The ghrelin receptor (GHS-R) is a seven transmembrane (7-TM) GPCR that is expressed in the hippocampus, pituitary, and hypothalamus, as well as in peripheral tissues including the heart, lung, liver, kidney, pancreas, intestines, and immune cells. Ghrelin signals are primarily relayed via coupling of the GHS-R to Gq/11 alpha subunits, promoting stimulation of phospholipase C, and an increase in intracellular calcium. Stimulation of adenylyl cyclases and increased cAMP are also induced by GHS-R activation. In humans and swine, two splice variants of the ghrelin receptor are expressed from the same gene. The GHS-R 1(a) receptor is the 366 amino acid, 7-TM form, while GHS-R 1(b) is a 289 amino acid, 5-TM form that is truncated at the C-terminus and is pharmacologically inactive. The epitope recognized by this antibody is unaffected by alternative splicing.

Recognizes rat ghrelin receptor. Predicted MW of ~41kDa. Higher MW species (~75kDa) and intermediates also observed by Western blot which reflect post-translational modification and heterodimerization of the receptor. Predicted species reactivity based on epitope sequence homology: human, mouse, marmoset, rhesus macaque, platypus, horse.

For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Informations de commande: Manufactured by Proteimax.

Type: Primary
Antigen: GHRELIN RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité