Anti-HOP Mouse Monoclonal Antibody
Fournisseur: ENZO LIFE SCIENCES
ADISRA1500D ADISRA1500F
ENZOADISRA1500DEA 290 EUR InStock
ENZOADISRA1500D ENZOADISRA1500F
Anti-HOP Mouse Monoclonal Antibody
Anticorps
Hsp70-Hsp90 Organizing Protein (HOP, p60) is an ~60kDa protein that is a critical intermediate component for the efficient maturation of steroid receptor complexes, serving to recruit Hsp90 to Hsp70-containing complexes. HOP contains three tetratricopeptide repeat (TPR) domains, TPR1, TPR2a, and TPR2b.
  • For Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, mink, pig, rabbit, sheep and Xenopus HOP. Detects a band of ~60kDa by Western blot.
Type: Primary
Antigen: HOP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité