Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-BSEP Mouse Monoclonal Antibody

Fournisseur: ENZO LIFE SCIENCES AG
ALEXALX801035C100EA 539 EUR InStock
ALEXALX801035C100 ENZOALX801035C100
Anti-BSEP Mouse Monoclonal Antibody
Anticorps
Bile Salts
Bile salts are important physiological agents that serve a number of functions, including absorption, solubilization, transport and secretion of lipids. In the liver, they participate in the generation of bile flow and the secretion of cholesterol and phospholipids, such as phosphatidylcholine. When released into the intestine, they facilitate the uptake of cholesterol, fat-soluble vitamins and other lipids. Moreover, the biosynthesis of bile acids from cholesterol is the most significant pathway for the elimination of cholesterol from the body. However, because of their detergent properties, bile acids are inherently cytotoxic and disruptions in their normal transport or secretion can result in a variety of pathophysiological conditions.

BSEP
Bile formation is an important function of the liver. It is mediated by hepatocytes which generate bile flow within the bile canaliculi by continuous vectorial secretion of bile salts and other solutes across their canalicular (apical) membrane. Bile secretion is mediated by several ATP-binding cassette (ABC) transporters located in the canalicular membrane of hepatocytes. Among these ABC transporters, the bile salt export pump (BSEP or ABCB11) represents the primary, if not sole transport system for the canalicular excretion of bile salts. Bile secretory failure results in cholestasis and progressive familial intrahepatic cholestasis (PFIC) in infancy represents a group of inherited cholestatic diseases that are classified into three subtypes. One of these subtypes, PFIC II, is associated with mutations in the BSEP gene. PFIC patients with mutations in the BSEP gene have normal γ-glutamyltransferase activity, low concentrations of bile salts in bile, and an absence of bile duct proliferation. Additionally, human obesity is associated with altered cholesterol homeostasis including increased production and turnover, as well as secretion of excess cholesterol from the liver into bile.

BSEP is a multifunctional polypeptide with two homologous halves, each containing a hydrophobic membrane-ancoring domain and an ATP-binding cassette (ABC) domain. The membrane-anchoring domain is composed of six helixes buried in the lipid bilayer of the plasma membrane and the ATP binding ABCs are exposed to the cytosol. The membrane-anchoring domain helixes are thought to form channels spanning the plasma membrane.

  • For Immunocytochemistry, Western Blot

Recognizes human BSEP. Does not cross-react with human P-glycoprotein.

Type: Primary
Antigen: BSEP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité