PBS concentrate
Fournisseur: Enzo Life Sciences
ENZOADI801927EA 61.6 EUR InStock
ENZOADI801927 ALEXADI-80-1927
PBS concentrate
Tampons Tampons d'électrophorèse Tampons bloquants
This PBS concentrate is a general all-purpose buffer specifically designed for use with the ImmunoSet products. Each bottle is sufficient for use with 5x96 wells microtiter plates if following the instructions in our ImmunoSet protocol.
The PBS concentrate may be diluted and easily modified by the addition of BSA and Tween®-20, as appropriate, to create the plate coating buffer, blocking buffer, and wash solution, in addition to creating the standard and sample diluent. Solutions of BSA and Tween are also available (Prod. No. ASSAADI-80-1928 and ASSAADI-80-1929, respectively). Additional assay development reagents are available as companion products to our ImmunoSet products.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité