TXB2 ELISA Kits
Fournisseur: Enzo Life Sciences
ALEXADI-901-002EA 2500 EUR InStock
ALEXADI-901-002 ENZOADI901002
TXB2 ELISA Kits
Tests Tests ELISA
For the quantitative determination of TXB2 in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate, coronary effluent, liver perfusate, platelets, rectal dialysate, and whole blood.
The TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 in a convenient 96 well plate based assay. This kit is highly cited in peer-reviewed publications in a diverse assortment of sample types including mucosa, tissue, serum, plasma, urine, and much more. The TXB2 EIA kit provides easy-to-use stable liquid color-coded reagents that reduce errors.
Informations de commande: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-054 (1x96 wells).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité