Testosterone ELISA kit
Fournisseur: Enzo Life Sciences
ALEXADI-900-065EA 530 EUR InStock
ALEXADI-900-065 ENZOADI900065
Testosterone ELISA kit
Tests Tests ELISA
For the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species. Not suitable for human serum and plasma. For those samples use product 89133-298. Cited sample types include tissue and feces.
The Testosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. Ready-to-use liquid color-coded reagents are provided to reduce error. The reproducible results lot-after-lot provide excellent consistency, ideal for longitudinal studies.
Informations de commande: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer 3, Wash buffer concentrate, Standard, Steroid displacement reagent, pNpp Substrate, Stop solution. Alternative size available: 89141-308 (5x96 wells)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité