AMPIGENE® qPCR Green Mix Hi-ROX
Fournisseur: ENZO LIFE SCIENCES
ENZNUC1040200 ENZNUC1041000
ENZOENZNUC1040200EA 124.3 EUR InStock
ENZOENZNUC1040200 ENZOENZNUC1041000
AMPIGENE® qPCR Green Mix Hi-ROX
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
Optimised dye-based qPCR.
AMPIGENE® qPCR Green Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing dye-based real-time PCR with minimal optimization. AMPIGENE® qPCR Green Mixes use a proprietary intercalating dye that does not inhibit PCR.An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Green Mixes come with either Lo-ROX or Hi-ROX concentration. For Lo-ROX, see Prod. No. ENZ-NUC103. Choose the appropriate ROX concentration for the instrument.
Informations de commande: Contains 2x AMPIGENE® qPCR Green Mix Hi-ROX (includes dNTPs and polymerases).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité