AMPIGENE® qPCR Probe Mix No-ROX
Fournisseur: ENZO LIFE SCIENCES
ENZNUC1070200 ENZNUC1071000
ENZOENZNUC1070200EA 209 EUR InStock
ENZOENZNUC1070200 ENZOENZNUC1071000
AMPIGENE® qPCR Probe Mix No-ROX
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
Optimised probe-based qPCR.
AMPIGENE® qPCR Probe Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing probe-based real-time PCR with minimal optimization. AMPIGENE® qPCR Probe Mixes are designed for use on a variety of probes including: Taqman ®, Molecular Beacons ®, and Scorpion probes ®. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Probe Mixes come with either Lo-ROX, Hi-ROX, or No-ROX concentration. For Lo-ROX and Hi-ROX, see Prod. Nos. ENZ-NUC105 and ENZ-NUC106. Choose the appropriate ROX concentration for the instrument (see instrument compatibility for reference dye chart below.)
Informations de commande: Contains 2x AMPIGENE® qPCR Probe Mix NO-ROX (includes dNTPs and polymerases).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité