Abz-SDK(Dnp)-P
Fournisseur: Enzo Life Sciences
ENZOBMLP1620001EA 137.5 EUR InStock
ENZOBMLP1620001 ALEXBML-P162-0001
Abz-SDK(Dnp)-P
Protéines et Peptides
ACE substrate
Fluorogenic substrate for angiotensin converting enzyme-1 (ACE1) N-domain. The somatic form of ACE1 is a zinc-dependent peptidyl dipeptidase comprising two catalytic domains referred to as the N- and C-domains. The testicular form, due to a downstream start site, contains only the C-domain. Abz fluorescence is quenched by the Dnp group until cleavage (at Asp-Lys) separates them. Ex: 320 nm, Em: 420 nm. This quenched substrate is useful for assay of cell culture ACE1 N-domain activity, inhibitor screening and kinetic analysis.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité