Human Recombinant HDAC10 (from Insect cells (Sf9)), HIS Tag
Fournisseur: Enzo Life Sciences
ENZOBMLSE5590050EA 578.6 EUR InStock
ENZOBMLSE5590050 ALEXBML-SE559-0050
Human Recombinant HDAC10 (from Insect cells (Sf9)), HIS Tag
Protéines et Peptides
Highly active
Produced in Sf9 cells. HDAC10 from human cDNA, transcript variant 1 (identical to GenBank accession NM_032019) is fused at the C-terminus to a His-tag.
Deacetylates lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité