Caspase inhibitor set III
Fournisseur: Enzo Life Sciences
ENZOALX850227KI01EA 765.6 EUR InStock
ENZOALX850227KI01 ALEXALX850227KI01
Caspase inhibitor set III
Protéines et Peptides
Synthetic peptides that irreversibly inhibit the activity of each of the caspase-family proteases. YVAD is the recognition sequence for caspase-1/ICE, VDVAD is the recognition sequence for caspase-2, DEVD is the recognition sequence for caspase-3/CPP32, WEHD is the recognition sequence for caspase-5, VEID is the recognition sequence for caspase-6/Mch2, IETD is the recognition sequence for caspase-8/FLICE, LEHD is the recognition sequence for caspase-9, and VAD is the recognition sequence for a caspase(s) that functions upstream of most other caspases.
Informations de commande: Contents: Contains 25 µl (2 mM) each of the following inhibitors:
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-2 Inhibitor, Z-VDVAD-FMK, orange cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-5 Inhibitor, Z-WEHD-FMK, clear cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-9 Inhibitor, Z-LEHD-FMK, blue cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité