Human catestatin
Fournisseur: Enzo Life Sciences
ALEXBMLCP64200001EA 385 EUR InStock
ALEXBMLCP64200001 ENZOBMLCP64200001
Human catestatin
Protéines et Peptides
Human Chromogranin A [352-372]). Wild type catestatin, initially described as a potent catecholamine release inhibitory peptide. Cst located in the skin is active against Gram-negative and Gram-positive bacteria as well as against fungi and yeast. Also proposed as a modulator or regulator of cardiac function and as an anti-hypertensive agent.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité