36415.22
36415.36
36415.A3
36415.22EA
33.3
EUR
InStock
36415.22
36415.36
36415.A3
Acide lactique ≥85.0-90.0% ACS
Acide lactique
CAS No.: 50-21-5
Molecular Formula: C3H6O3
Formula Weight: 90.08
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 1.209
Refractive Index: 1.4270
Appearance: Colourless to yellow
Melting Point: 18°
Boiling Point: 122°/15mm
Beilstein Reference: 1209341
Merck Reference: 14,5336
MDL No.: MFCD00004520