ESD safety ankle boots, lace-up, uvex motion light, 6986
Fournisseur: UVEX
6986.2/38 6986.2/39 6986.2/40 6986.2/41 6986.2/42 6986.2/43 6986.2/45 6986.2/46 6986.2/47 6986.2/48
UVEX6986.2/38EA 118 EUR InStock
UVEX6986.2/38 UVEX6986.2/39 UVEX6986.2/40 UVEX6986.2/41 UVEX6986.2/42 UVEX6986.2/43 UVEX6986.2/45 UVEX6986.2/46 UVEX6986.2/47 UVEX6986.2/48
ESD safety ankle boots, lace-up, uvex motion light, 6986
Chaussures et bottes
Ultralight ESD ankle boots made from full-grain leather, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. +120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Ergonomically designed, self-cleaning tread
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Width: 11
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, SRC
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité