B24901.06
B24901.14
B24901.06EA
72.6
EUR
InStock
B24901.06
B24901.14
2-Bromo-5-fluoroaniline ≥98%
2-Bromo-5-fluoroaniline
Purity: 98%
CAS No.: 1003-99-2
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Flash Point: 110°(230°F)
Melting Point: 42-46°
Boiling Point: 154°/23mm
UN No.: 2811
MDL No.: MFCD00070750