Synonyms:
3,4-Dicyanoaniline, 4-Aminophthalonitrile
L13433.03
L13433.06
L13433.03EA
60.5
EUR
InStock
L13433.03
L13433.06
3,4-Dicyanoaniline 98%
3,4-Dicyanoaniline
Purity: 98%
CAS No.: 56765-79-8
Molecular Formula: C8H5N3
Formula Weight: 143.15
Melting Point: 179-182°
Beilstein Reference: 2937382
UN No.: 3439
MDL No.: MFCD00041741