L20124.06
L20124.14
L20124.06EA
75.4
EUR
InStock
L20124.06
L20124.14
3-Éthynylaniline 98%
3-Éthynylaniline
Purity: 98%
CAS No.: 54060-30-9
Molecular Formula: C8H7N
Formula Weight: 117.15
Flash Point: 100°(212°F)
Refractive Index: 1.6170
Melting Point: 27°
Boiling Point: 70°/1mm
Beilstein Reference: 2935417
MDL No.: MFCD00014779