ESD safety shoes, lace-up, uvex 1, 8531
Fournisseur: UVEX
85319/46 85319/42 85319/43 85319/48 85319/49 85319/47 85319/50 85319/51 85319/52 85319/38 85319/39 85319/35 85319/36 85319/37 85319/40 85319/41
UVEX85319/46EA 0 EUR InStock
UVEX85319/46 UVEX85319/42 UVEX85319/43 UVEX85319/48 UVEX85319/49 UVEX85319/47 UVEX85319/50 UVEX85319/51 UVEX85319/52 UVEX85319/38 UVEX85319/39 UVEX85319/35 UVEX85319/36 UVEX85319/37 UVEX85319/40 UVEX85319/41
ESD safety shoes, lace-up, uvex 1, 8531
Chaussures et bottes
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/blue
Width: 12
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité