L03477.06
L03477.14
L03477.06EA
59.6
EUR
InStock
L03477.06
L03477.14
Chlorotributylsilane ≥97%
Chlorotributylsilane
Purity: 97%
CAS No.: 995-45-9
Molecular Formula: C12H27ClSi
Formula Weight: 234.89
Flash Point: 104°(219°F)
Refractive Index: 1.4460
Boiling Point: 93-94°/4mm
Beilstein Reference: 1742537
UN No.: 2987
MDL No.: MFCD00000509