Safety Shoes, sandals, Flame, Harvey Easy, Jori
Fournisseur: ELTEN GMBH
11721/36 11721/37 11721/38 11721/39 11721/40 11721/41 11721/42 11721/43 11721/44 11721/45 11721/46 11721/47 11721/48
ELTN11721/36EA 61.9 EUR InStock
ELTN11721/36 ELTN11721/37 ELTN11721/38 ELTN11721/39 ELTN11721/40 ELTN11721/41 ELTN11721/42 ELTN11721/43 ELTN11721/44 ELTN11721/45 ELTN11721/46 ELTN11721/47 ELTN11721/48
Safety Shoes, sandals, Flame, Harvey Easy, Jori
Chaussures et bottes
Metal-free safety sandals made from breathable suede combined with quick drying tear resistant textile, skin friendly textile lining, a full padded bellows tongue, collar padding, plus a dual-density PU/rubber Flame sole (4,0 mm profile, oil, fuel and chemical resistant, also temperature resistant from approx. –20 and +200 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • PU toe protection cap
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Blue/yellow
Certifications: EN ISO 20345; S1P, SRC.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité