Safety shoes, lace-up, Flame, Harvey Low, Jori
Fournisseur: ELTEN
12721/36 12721/37 12721/38 12721/39 12721/40 12721/41 12721/42 12721/43 12721/44 12721/45 12721/46 12721/47 12721/48
ELTN12721/36EA 61.9 EUR InStock
ELTN12721/36 ELTN12721/37 ELTN12721/38 ELTN12721/39 ELTN12721/40 ELTN12721/41 ELTN12721/42 ELTN12721/43 ELTN12721/44 ELTN12721/45 ELTN12721/46 ELTN12721/47 ELTN12721/48
Safety shoes, lace-up, Flame, Harvey Low, Jori
Chaussures et bottes
Metal-free safety shoes made from hydrophobised suede, combined with hydrophobised, quick drying, tear resistant textile, skin friendly textile lining, a full padded bellows tongue, collar padding, plus a dual-density PU/rubber Flame sole (4,0 mm profile, oil, fuel and chemical resistant, also temperature resistant from approx. –20 to +200 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • PU scuff cap protects the upper material
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects as well as areas close to induction loops or metal detectors.
Colour: Blue/yellow
Certifications: EN ISO 20345; S3, SRC.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité