Safety shoes, lace-up, Flame, Harrison Low, Jori
Fournisseur: ELTEN GMBH
12741/41 12741/42 12741/43 12741/44 12741/45 12741/46 12741/47 12741/48 12741/36 12741/37 12741/38 12741/39 12741/40
ELTN12741/41EA 60.9 EUR InStock
ELTN12741/41 ELTN12741/42 ELTN12741/43 ELTN12741/44 ELTN12741/45 ELTN12741/46 ELTN12741/47 ELTN12741/48 ELTN12741/36 ELTN12741/37 ELTN12741/38 ELTN12741/39 ELTN12741/40
Safety shoes, lace-up, Flame, Harrison Low, Jori
Chaussures et bottes
Metal-free safety shoes made from hydrophobised suede, skin-friendly textile lining, a full padded bellows tongue, collar padding, and a double density PU/rubber FLAME sole (4,0 mm profile, oil, fuel and chemical resistant, temperature resistant from approx. -20 and +200 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • PU scuff cap protects the upper material
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects as well as areas close to induction loops or metal detectors.
Colour: Black/yellow
Certifications: EN ISO 20345; S3, SRC
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité