Safety ankle boots, lace-up, Basic, Achim S2, Jori
Fournisseur: ELTEN GMBH
16320/36 16320/37 16320/38 16320/39 16320/40 16320/41 16320/42 16320/43 16320/44 16320/45 16320/46 16320/47 16320/48
ELTN16320/36EA 39.9 EUR InStock
ELTN16320/36 ELTN16320/37 ELTN16320/38 ELTN16320/39 ELTN16320/40 ELTN16320/41 ELTN16320/42 ELTN16320/43 ELTN16320/44 ELTN16320/45 ELTN16320/46 ELTN16320/47 ELTN16320/48
Safety ankle boots, lace-up, Basic, Achim S2, Jori
Chaussures et bottes
Durable safety ankle boots made from wear resistant cowhide leather, skin friendly textile lining, a full padded bellows tongue and collar padding, plus a dual-density PU/PU New Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
Certifications: EN ISO 20345; S2, SRC.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité