Safety ankle boots, lace-up, Flame, Harvey Mid, Jori
Fournisseur: ELTEN GMBH
16721/36 16721/37 16721/38 16721/39 16721/40 16721/41 16721/42 16721/43 16721/44 16721/45 16721/46 16721/47 16721/48
ELTN16721/36EA 62.9 EUR InStock
ELTN16721/36 ELTN16721/37 ELTN16721/38 ELTN16721/39 ELTN16721/40 ELTN16721/41 ELTN16721/42 ELTN16721/43 ELTN16721/44 ELTN16721/45 ELTN16721/46 ELTN16721/47 ELTN16721/48
Safety ankle boots, lace-up, Flame, Harvey Mid, Jori
Chaussures et bottes
Metal-free safety shoes made from hydrophobised suede combined with hydrophobised, quick drying tear resistant textile, skin friendly textile lining, a full padded bellows tongue, collar padding, plus a dual-density PU/rubber Flame sole (4,0 mm profile, oil, fuel and chemical resistant, also temperature resistant from approx. –20 and +200 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • PU scuff cap protects the upper material
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects as well as areas close to induction loops or metal detectors.
Colour: Blue/yellow
Certifications: EN ISO 20345; S3, SRC.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité