ESD safety shoes, lace-up, uvex motion light, 6982
Fournisseur: UVEX
6982.8/38 6982.8/39 6982.8/40 6982.8/41 6982.8/42 6982.8/43 6982.8/44 6982.8/45 6982.8/46 6982.8/47 6982.8/48
UVEX6982.8/38EA 92.9 EUR InStock
UVEX6982.8/38 UVEX6982.8/39 UVEX6982.8/40 UVEX6982.8/41 UVEX6982.8/42 UVEX6982.8/43 UVEX6982.8/44 UVEX6982.8/45 UVEX6982.8/46 UVEX6982.8/47 UVEX6982.8/48
ESD safety shoes, lace-up, uvex motion light, 6982
Chaussures et bottes
Wide, sports-fit ESD safety shoes made from nubuck leather upper, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. +120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Colour: Black
Width: 11
Certifications: EN 61340 (ESD); EN ISO 20345:2011; DGUV 112-191; S1, SRC
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité