Organic reference standard, neat, Dimethyl sulfone, (CRM), standard for quantitative NMR, TraceCERT®, Supelco®
Fournisseur: Merck
Supelco®
41867-1GEA 127 EUR InStock
41867-1G
Organic reference standard, neat, Dimethyl sulfone, (CRM), standard for quantitative NMR, TraceCERT®, Supelco®
Étalons Étalons organiques
Dimethyl sulfone has been used as internal standard, for the nuclear magnetic resonance spectrum studies of cocrystals containing API carbamazepine, synthesized using mechanochemical method of synthesis.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité