Alcohol Metabolite Standards, PEth 16:0/18:1-D5 solution (phosphatidylethanol 16:0/18:2), CRM, Cerilliant®, Supelco®
Fournisseur: Merck
Cerilliant®
P-117-1MLEA 360 EUR InStock
P-117-1ML
Alcohol Metabolite Standards, PEth 16:0/18:1-D5 solution (phosphatidylethanol 16:0/18:2), CRM, Cerilliant®, Supelco®
Étalons Étalons organiques
PEth 16:0/18:2, also known as phosphatidylethanol 16:0/18:2, is a sensitive and specific biomarker for alcohol use and abuse.
  • Snap-N-Spike®/Snap-N-Shoot®
Since the detection window of PEth 16:0/18:2, in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:2, levels is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:2 has also been investigated as a biomarker to confirm prenatal alcohol exposure (PAE).
Attention: Storage temperature: −70 °C (Ultra Low freezer).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité