52010.EA
92.4
EUR
InStock
52010.
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ) ≥95% (par HPLC) indicateur fluorescent, pour détermination du chlorure
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ)
SPQ is a blue fluorescent dye (Ex/Em 344/443 nm) that is specifically quenched by chloride via collision (KSV=118 M-1). Therefore, chloride concentrations can be measured by moniotiring the degree of fluorescence decrease. The dye can be loaded into cells by hypotonic shock.
- White solid soluble in H2O and DMSO
- Store at 4°C. Protect from light, especially in solution.
- λEx/λEm= 344nm / 443nm
- ε (344 nm, H2O) = 3,700