ESD safety shoes, lace-up, uvex 2 xenova®, 9560
Fournisseur: UVEX
9560.9/51
UVEX9560.9/51EA 123 EUR InStock
UVEX9560.9/51
ESD safety shoes, lace-up, uvex 2 xenova®, 9560
Chaussures et bottes
Very lightweight fashionable uvex 2 xenova® perforated S1 safety shoe made from synthetic materials, soft padded collar, suitable for people allergic to chrome. All sole materials are free of silicones, plasticisers and other paint-wetting impairment substances.
  • Compact design, anatomical shape, good lateral stability, no thermal conductivity
  • Climazone technology for improved comfort
  • Breathable materials as well as the perforated upper material
  • Removable anti-static comfortable insole, with moisture transport system
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Shock absorption around the heel and forefoot
  • Metal-free uvex xenova® toe cap
  • Fulfils the ESD requirement: Discharge resistance <35 megohm
Used for medium-duty applications.
Colour: Black/green
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité