Pesticide reference standard, Dithianon, PESTANAL®, analytical standard, Supelco®
Fournisseur: Merck
Supelco® PESTANAL®
45462-250MGEA 76.9 EUR InStock
45462-250MG
Pesticide reference standard, Dithianon, PESTANAL®, analytical standard, Supelco®
Étalons Étalons organiques
Dithianons belong to the quinone class of fungicides.
It is determined in various applications,
Surface water samples by solid-phase extraction (SPE) and liquid chromatography coupled to quadrupole time-of-flight mass spectrometry (LC-Q-TOFMS)
Honeybees by dispersive-SPE followed by liquid and gas chromatography coupled to tandem mass spectrometry (LC-MS/MS and GC-MS/MS) equipped with electrospray ionization (ESI) and multiple reaction monitoring (MRM) detection
Apples by QuEChERS extraction, high performance thin-layer chromatography (HPTLC) and GC-MS/MS
Tomatoes by QuEChERS extraction and LC coupled to triple quadrupole (QqQ) ESI-MS/MS with MRM detection
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité