Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95% (par HPLC) colorant pour mitochondries, convient pour la fluorescence
Fournisseur: Biotium
Attention
70016.EA 331 EUR InStock
70016.
Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95% (par HPLC) colorant pour mitochondries, convient pour la fluorescence
Tetramethylrhodamine ethyl ester perchlorate (TMRE)
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM (70017) are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation.
  • Cellular localization: Mitochondria
  • For live or fixed cells: For live/intact cells
  • Assay type/options: Real-time imaging
  • Cell permeability: Membrane permeant
  • Apoptosis/viability marker: Mitochondrial potential
  • Potential dependence: Mitochondrial potential-dependent
  • λEx/λEm (MeOH) = 549/574 nm
  • Red solid soluble in DMSO, DMF or EtOH
  • Store at 4°C and protect from light
TMRE (Tetramethylrhodamine ethyl ester, perchlorate) and TMRM (70017) are preferred dyes for quantitative measurements of membrane potentials using the Nernst equation. The dyes do not form aggregates in cell membranes and have minimal interaction with membrane proteins. Thus, the transmembrane distribution of the dyes is directly related to the membrane potential according to the Nernst equation. Biotium also offers TMRE, 2 mM in DMSO (70005) as a ready to use stock solution.
Formule: C₂₆H₂₇ClN₂O₇
MW: 514,95 g/mol
Numéro MDL: MFCD00467973
Numéro CAS: 115532-52-0
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité