Organic reference standard, (RS)-Mevalonic acid lithium salt, analytical standard, Supelco®
Fournisseur: Merck
Supelco®
42147-5MGEA 80.5 EUR InStock
42147-5MG
Organic reference standard, (RS)-Mevalonic acid lithium salt, analytical standard, Supelco®
Étalons Étalons organiques
Biochemical and physiol actions are metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone.
  • Analysis note is NMR-Internal standard: dimethylsulfone
Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.
Conditionnement: Bottomless glass bottle. Contents are inside inserted fused cone.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité