Organic reference standard, Dehydrocostus lactone, analytical standard, Supelco®
Fournisseur: Merck
Supelco®
42575-10MGEA 76.8 EUR InStock
42575-10MG
Organic reference standard, Dehydrocostus lactone, analytical standard, Supelco®
Étalons Étalons organiques
Suppressors of cytokine signaling (SOCS) proteins negatively regulate cytokine receptor signaling. The SOCS family include eight members SOCS-1 to SOCS-7 and CIS. Dehydrocostuslactone (DHE) is a sesquiterpene lactone extracted from Saussurea lappa and Aucklandia lappa. The compound possess antifungal and antimycobacterial activity and exhibit a cytotoxic effect against HepG2, OVCAR-3, and HeLa cell lines. DHE inhibits cell proliferation not only in early-stage breast cancer MCF-7 but also in more advanced stage MDA-MB-231 cancer cells, without significant effect on H184B5F5/M10 normal mammary epithelial cells. The inhibition of breast cancer cells proliferation occurs through cell cycle blockade and apoptosis. DHE inhibits cell cycle progression and cell death through SOCS-1 and SOCS-3.
Attention: P261 - P272 - P280 - P302 + P352 - P333 + P313 - P362 + P364
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité