Biotin-CF® 633 colorant fluorescent
Fournisseur: Biotium
80031.EA 325 EUR InStock
80031.
Biotin-CF® 633 colorant fluorescent
Biotin-CF®
Potential applications of Biotin-CF® dye include detection of biotin binding sites and the degree of biotinylation of proteins, and for the measurement of avidin and streptavidin in crude biological samples. In addition, biotin-CF® dye can be used as a polar tracer by microinjection to study the morphology of cells. CF® dyes are very photostable which make them suitable for studies where prolonged exposure to light may be necessary.
  • Ex/Em 630/650 nm
  • Dark Blue solid soluble in DMSO or DMF
CF® Dye Biotin is available with the following CF® dyes: CF®568, CF®633, and CF®640R.
Attention: Store desiccated at 4 °C. Protect from light.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité