Disposable particle filtering respirator, FFP3, X-plore® 1900
Fournisseur: Dräger
X-plore®
DRAE3951935EA 88.8 EUR InStock
DRAE3951935
Disposable particle filtering respirator, FFP3, X-plore® 1900
Respirateurs Masques à usage unique Demi-masques fitrants contre les particules
X-plore® 1900 is a high-performance, single-use dust mask for protection against fine dust and particles. The premium particle filtering mask is available in two sizes, combining ease of use, exceptional comfort, and optimal fit.
  • Can be worn with spectacles
  • Tear-resistant head straps
  • Flexible nose clip and soft nose pad
  • Soft, moisture-resistant inner lining
  • Three-part folding design
  • Available in two sizes (S or M/L)
This X-plore® 1900 folded particle filtering face piece provides effective protection against fine dust and particles. Protection class FFP3 (filter efficiency min. 99%). The mask offers optimal fit and seal, easy handling, high wearing comfort, and is compatible with protective eyewear.
CoolMAX exhalation valve effectively releases humid and warm exhaled air to the outside. Use for max. one work shift. Additional dolomite dust test passed. EAN number: 04026056118182.
Certifications: EN 149:2001+A1:2009.
Informations de commande: Minimum order quantity of 10.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité