Rat Recombinant Prolactin (from E. coli)
Fournisseur: Shenandoah Biotechnology
SHBT300-11AF-1MGEA 1540 EUR InStock
SHBT300-11AF-1MG SHBT300-11AF-500UG SHBT300-11AF-50UG SHBT300-11AF-100UG SHBT300-11AF-10UG
Rat Recombinant Prolactin (from E. coli)
Protéines et Peptides
Prolactin (PRL) is a hormone that is produced and secreted by the pituitary gland. It acts in an endocrine, paracrine, and autocrine manner. The prolactin receptor (PRLR) is expressed on many cell types, including cells of the reproductive organs, central nervous system, and breast cancer.
  • Animal-free
  • Produced from genetically modified E. coli
  • Contains 198 amino acids
PRL signal transduction occurs via JAK kinase signaling pathways. The primary function of PRL is to regulate lactation, but PRL also plays functional roles in the immune system and during cell growth, apoptosis, and differentiation.
Information de livraison: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Attention: For research use only and is not for use in humans.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité